Learn More
Medchemexpress LLC MEK4 inhibitor-1 | 2570386-36-4 | 98.9% | 291.30 | 50 MG
SDP
Catalog No. 5000349290
Encompass_Preferred
MEK4 inhibitor-1 is a novel MEK4 inhibitor developed for its activity against pancreatic adenocarcinoma.
  • Novel MEK4 inhibitor
  • Active against pancreatic adenocarcinoma
  • IC50 value of 61 nM
Catalog No. 50-003-49290 Supplier Medchemexpress LLC Supplier No. HY13963850MG
MEK4 inhibitor-1 is a novel MEK4 inhibitor developed for its activity against pancreatic adenocarcinoma.
  • Novel MEK4 inhibitor
  • Active against pancreatic adenocarcinoma
  • IC50 value of 61 nM