Learn More
Enzo Life Sciences MEKK 3 polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ADIKASMA013E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of mouse MEKK3. Source: Whole rabbit serum. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Drosophila, Dog. Applications: IHC (PS), WB. Formulation: Liquid. Whole rabbit serum. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7316