Learn More
Hach Company MEL/MF Total Coliform Laboratory
Supplier:  Hach Company 2569700
Encompass
The Total Coliform MEL Laboratory is designed for analysts who need portable or small-scale microbiology testing capabilities. From a remote field location to a small scale microbiology lab within your existing lab, this MEL offers all the equipment and accessories to perform microbiological analysis.Contains: Portable Incubator and rack, Thermometer, Whirlpak bags with thiosulfate (200), Filter funnel/pad assemblies (200), alcohol burner (order alcohol separately), vacuum filtration apparatus, colony counter, and permanent marker.Choose and order media separately.
Catalog No. 50-199-6252
Add to Cart
Request a Quote