Learn More
Medchemexpress LLC MEL24 | 642072-48-8 | C17H18N4O3 | 25 MG
SDP
Catalog No. 5000374678
Encompass_Preferred
MEL24 is an Mdm2 E3 ligase inhibitor that reduces cell survival and increases sensitivity to DNA damaging agents in a p53-dependent manner for in vitro antitumor studies.
  • Reduces cell survival
  • Increases sensitivity to DNA damaging agents
  • Suitable for in vitro antitumor studies
  • Acts on 293T cells with an ED50 value of 9.2 μM
  • Increases levels of Mdm2, p53, and MdmX
  • Activates p53
  • Inhibits the degradation of endogenous MdmX in HCT116 cells
Catalog No. 50-003-74678 Supplier Medchemexpress LLC Supplier No. 25MGHY153215
MEL24 is an Mdm2 E3 ligase inhibitor that reduces cell survival and increases sensitivity to DNA damaging agents in a p53-dependent manner for in vitro antitumor studies.
  • Reduces cell survival
  • Increases sensitivity to DNA damaging agents
  • Suitable for in vitro antitumor studies
  • Acts on 293T cells with an ED50 value of 9.2 μM
  • Increases levels of Mdm2, p53, and MdmX
  • Activates p53
  • Inhibits the degradation of endogenous MdmX in HCT116 cells