Learn More
Medchemexpress LLC Melagatran | 159776-70-2 | 99.3% | 1 MG
SDP
Catalog No. 5000341139
Encompass_Preferred
Melagatran is a reversible, selective, and orally active direct inhibitor of thrombin, with a Ki of 2 nM. It functions by binding directly to the active site of thrombin, inhibiting the conversion of fibrinogen to fibrin. This compound also reduces the DNA binding activity of NF-κB and AP-1, making it relevant for research in cardiovascular diseases and ischemic brain damage. It has been shown to alleviate ischemic brain damage and decrease the size of advanced atherosclerotic lesions.
  • Targets thrombin with a Ki of 2 nM.
  • Alleviates endotoxin-induced systemic hemodynamic disorders.
  • Prevents or delays electrically induced occlusive thrombus formation.
  • Demonstrates good oral bioavailability.
  • Reduces ischemic brain injury in rats.
  • Reduces advanced atherosclerotic lesion size and promotes plaque stability.
  • Decreases DNA binding activity of NF-κB and AP-1.
Catalog No. 50-003-41139 Supplier Medchemexpress LLC Supplier No. HY1290561MG
Melagatran is a reversible, selective, and orally active direct inhibitor of thrombin, with a Ki of 2 nM. It functions by binding directly to the active site of thrombin, inhibiting the conversion of fibrinogen to fibrin. This compound also reduces the DNA binding activity of NF-κB and AP-1, making it relevant for research in cardiovascular diseases and ischemic brain damage. It has been shown to alleviate ischemic brain damage and decrease the size of advanced atherosclerotic lesions.
  • Targets thrombin with a Ki of 2 nM.
  • Alleviates endotoxin-induced systemic hemodynamic disorders.
  • Prevents or delays electrically induced occlusive thrombus formation.
  • Demonstrates good oral bioavailability.
  • Reduces ischemic brain injury in rats.
  • Reduces advanced atherosclerotic lesion size and promotes plaque stability.
  • Decreases DNA binding activity of NF-κB and AP-1.