Learn More
Medchemexpress LLC Melagatran | 159776-70-2 | 99.3% | 10 MG
SDP
Catalog No. 5000341335
Encompass_Preferred
Melagatran is a reversible, selective, orally active direct inhibitor of thrombin with a Ki of 2 nM. It directly binds to the active site of thrombin, inhibiting the thrombin-mediated conversion of fibrinogen to fibrin. Melagatran reduces the DNA binding activity of NF-κB and AP-1, and has shown to reduce fibrin deposition in organs, alleviate ischemic brain damage, and reduce the size of advanced atherosclerotic lesions.
  • Reversible, selective thrombin inhibitor
  • Inhibits thrombin-mediated fibrinogen to fibrin conversion
  • Reduces DNA binding activity of NF-κB and AP-1
  • Alleviates ischemic brain damage
  • Decreases advanced atherosclerotic lesion size
  • Used in cardiovascular disease research
  • Investigated for neurological disease studies
Catalog No. 50-003-41335 Supplier Medchemexpress LLC Supplier No. HY12905610MG
Melagatran is a reversible, selective, orally active direct inhibitor of thrombin with a Ki of 2 nM. It directly binds to the active site of thrombin, inhibiting the thrombin-mediated conversion of fibrinogen to fibrin. Melagatran reduces the DNA binding activity of NF-κB and AP-1, and has shown to reduce fibrin deposition in organs, alleviate ischemic brain damage, and reduce the size of advanced atherosclerotic lesions.
  • Reversible, selective thrombin inhibitor
  • Inhibits thrombin-mediated fibrinogen to fibrin conversion
  • Reduces DNA binding activity of NF-κB and AP-1
  • Alleviates ischemic brain damage
  • Decreases advanced atherosclerotic lesion size
  • Used in cardiovascular disease research
  • Investigated for neurological disease studies