Learn More
Medchemexpress LLC Melamine | 108-78-1 | MFCD00006055 | 126.12 g/mol | C3H6N6 | 1000g
SDP
Catalog No. 5000268116
Encompass_Preferred
Melamine is an orally active inducer of Apoptosis Melamine induces animal disease models Melamine affects the activity of Sertoli cell and can be used for research on male reproductive function Melamine also has neurotoxicity and nephrotoxicity Melamine induces cognitive impairment and acute kidney injury models Melamine can also be used to induce bladder cancer and urinary stone models[1][2][3][4][5][6]
Catalog No. 50-002-68116 Supplier Medchemexpress LLC Supplier No. HYY11171000G
Melamine is an orally active inducer of Apoptosis Melamine induces animal disease models Melamine affects the activity of Sertoli cell and can be used for research on male reproductive function Melamine also has neurotoxicity and nephrotoxicity Melamine induces cognitive impairment and acute kidney injury models Melamine can also be used to induce bladder cancer and urinary stone models[1][2][3][4][5][6]