Learn More
Medchemexpress LLC Melamine (2,4,6-triamino-1,3,5-triazine) | 108-78-1 | MFCD00006055 | 99.9% | 126.12 g/mol | C3H6N6 | 250mg
SDP
Catalog No. 5000267133
Encompass_Preferred
Melamine (Standard) is the analytical standard of Melamine This product is intended for research and analytical applications Melamine is a metabolite of cyromazine Melamine is a intermediate for the synthesis of melamine resin and plastic materials[1]
Catalog No. 50-002-67133 Supplier Medchemexpress LLC Supplier No. HYY1117R250MG
Melamine (Standard) is the analytical standard of Melamine This product is intended for research and analytical applications Melamine is a metabolite of cyromazine Melamine is a intermediate for the synthesis of melamine resin and plastic materials[1]