Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melamine-(triamine-15N3), ≥95.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013828 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3828 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3828 Supplier MilliporeSigma Supelco Supplier No. 8003810MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 287476-11-3
Flash Point 300°C
Physical Form Neat
Linear Formula C3H615N3N3
MDL Number MFCD00190531
Melting Point >300°C
Synonym 1,3,5-Triazine-2,4,6-triamine-15N3
Type Analytical Standard
Quantity 10 mg
Formula Weight 129.10
Percent Purity ≥95.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.