Learn More
Biogenex Laboratories Melan-A : A103 : 100 test
SDP
Supplier:  Biogenex Laboratories AM36110M
Encompass_Preferred
Melan-A, product of MART-1 gene, is a differentiation antigen which is expressed in 100% of melanocytes, most melanomas, and 50-60% of melanoma cell lines. It is one of the melanoma antigens recognized by autologous cytotoxic T cells, and as an antigenic target for tumor infiltrating lymphocytes. This antibody also stains Melan-A in normal melanocytes and in retina. It does not stain normal or tumor tissues from nonmelanocyte lineages.
Catalog No. 50-197-8763