Learn More
Newcomer Supply Melan-A (MART-1) Control Slides, 10/set
SDP
Supplier:  Newcomer Supply 3580A
Encompass
Newcomer Supply Melan-A (MART-1) Control Slides are for the positive immunohistochemical staining of Melan-A, expressed in melanocytes of normal skin, retina, nevi, and in the vast majority of melanomas, and is useful in establishing the diagnosis of metastatic melanomas. Melan-A (MART-1) Control Slides contain sections of positive staining melanoma and negative staining lung.
Catalog No. NC2101947