Learn More
StatLab Melan-A Melanoma IHC Control Slides
SDP
Catalog No. 21010278
Change view
Click to view available options
Product Type:
Control Slide
Catalog No. Product Type
21-010-278 Control Slide
1 options
Catalog No. 21-010-278 Supplier StatLab Supplier No. CSMELANA/15
Add to Cart
Add to Cart
It consists of formalin fixed paraffin embedded tissue sections which contain known distinctive tissue elements.
  • One slide in each box features a serial section that has been stained using the specified staining procedure that is included in each box
  • All boxes contain 15 slides
Specifications
Product Type Control Slide
Quantity 15/Pk.