Learn More
Medchemexpress LLC HY-P1525A 1mg Medchemexpress, Melanin Concentrating Hormone, salmon (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1525A1MG
Encompass_Preferred
Medchemexpress, HY-P1525A 1mg Melanin Concentrating Hormone, salmon (TFA) CAS: Melanin Concentrating Hormone, salmon TFA (MCH (salmon) TFA) is a 19-amino-acid neuropeptide initially identified in the pituitary gland of teleost fish, which regulates food intake, energy balance, sleep state, and the cardiovascular system. Melanin-concentrating hormone is a ligand for an orphan G protein-coupled receptor (SLC-1/GPR24) and MCHR2 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4221