Learn More
StatLab Melanin (Human) Control Slides
SDP
Catalog No. 21010277
Change view
Click to view available options
Product Type:
Control Slide
Catalog No. Product Type
21-010-277 Control Slide
1 options
Catalog No. 21-010-277 Supplier StatLab Supplier No. CSMELA/25
Add to Cart
Add to Cart
It consists of formalin fixed paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratoies
  • StatLab control slides are created using Millennia adhesion white glass slides with 90° corners
  • Each package includes specific staining protocol
  • 25/Pk that includes 1 stained slide
  • Tissue - Human known positive
Specifications
Product Type Control Slide
Quantity 25/Pk.