Learn More
Medchemexpress LLC Melanin probe-1 | 1420844-62-7 | 98.1% | 239.29 | 50 MG
SDP
Catalog No. 5000351352
Encompass_Preferred
Melanin probe-1 is an 18F-picolinamides based PET probe designed for the PET imaging of malignant melanoma. It demonstrates high tumor targeting efficiency, excellent tumor imaging contrasts, desirable biodistribution patterns, and good in vivo stability.
  • Pet probe
  • Melanoma imaging
  • High tumor targeting
  • Excellent imaging contrast
  • Desirable biodistribution
  • Good in vivo stability
  • High selectivity and specificity
Catalog No. 50-003-51352 Supplier Medchemexpress LLC Supplier No. HY13640450MG
Melanin probe-1 is an 18F-picolinamides based PET probe designed for the PET imaging of malignant melanoma. It demonstrates high tumor targeting efficiency, excellent tumor imaging contrasts, desirable biodistribution patterns, and good in vivo stability.
  • Pet probe
  • Melanoma imaging
  • High tumor targeting
  • Excellent imaging contrast
  • Desirable biodistribution
  • Good in vivo stability
  • High selectivity and specificity