Learn More
Medchemexpress LLC Melanin probe-1 | 1420844-62-7 | MFCD32693924 | 98.1% | C12H18FN3O | 10MG
SDP
Catalog No. 5000260127
Encompass_Preferred
Melanin probe-1 is an 18F-picolinamide-based PET imaging probe developed for research use to image malignant melanoma. It exhibits high tumor targeting efficiency, strong tumor-to-background contrast, favorable biodistribution, and good in vivo stability, making it suitable for preclinical PET studies.
  • Designed for PET imaging of malignant melanoma.
  • High tumor targeting efficiency and strong contrast.
  • Favorable biodistribution and pharmacokinetics in vivo.
  • Good stability under biological conditions.
  • Provided for research use only.
Catalog No. 50-002-60127 Supplier Medchemexpress LLC Supplier No. HY13640410MG
Melanin probe-1 is an 18F-picolinamide-based PET imaging probe developed for research use to image malignant melanoma. It exhibits high tumor targeting efficiency, strong tumor-to-background contrast, favorable biodistribution, and good in vivo stability, making it suitable for preclinical PET studies.
  • Designed for PET imaging of malignant melanoma.
  • High tumor targeting efficiency and strong contrast.
  • Favorable biodistribution and pharmacokinetics in vivo.
  • Good stability under biological conditions.
  • Provided for research use only.