Learn More
Medchemexpress LLC Melanin probe-2 | 1285446-04-9 | 99.4% | 300.19 | 250 MG
SDP
Catalog No. 5000348783
Encompass_Preferred
Melanin probe-2 is a non-radioactive bromopicolinamide precursor. It is primarily used for the synthesis of 18F-Labeled Picolinamide PET probes.
  • Non-radioactive bromopicolinamide precursor.
  • Used for the synthesis of 18F-Labeled Picolinamide PET probes.
Catalog No. 50-003-48783 Supplier Medchemexpress LLC Supplier No. 250MGHY136405
Melanin probe-2 is a non-radioactive bromopicolinamide precursor. It is primarily used for the synthesis of 18F-Labeled Picolinamide PET probes.
  • Non-radioactive bromopicolinamide precursor.
  • Used for the synthesis of 18F-Labeled Picolinamide PET probes.