Learn More
Medchemexpress LLC Melanin probe-2 (compound 5) | 1285446-04-9 | 99.4% | 300.19 | 50 MG
SDP
Catalog No. 5000348207
Encompass_Preferred
Melanin probe-2 (compound 5) is a non-radioactive bromopicolinamide precursor. It is suitable for the synthesis of 18F-Labeled Picolinamide PET probe (HY-136404).
  • Non-radioactive bromopicolinamide precursor
  • Used for the synthesis of 18F-Labeled Picolinamide PET probe
Catalog No. 50-003-48207 Supplier Medchemexpress LLC Supplier No. HY13640550MG
Melanin probe-2 (compound 5) is a non-radioactive bromopicolinamide precursor. It is suitable for the synthesis of 18F-Labeled Picolinamide PET probe (HY-136404).
  • Non-radioactive bromopicolinamide precursor
  • Used for the synthesis of 18F-Labeled Picolinamide PET probe