Learn More
Promocell GmbH PromoCell Melanocyte Growth Medium 500 ml, ready-to-use, serum-free, with C-39415 attached to the flask
Catalog No. 50306230
Encompass
PromoCell Melanocyte Growth Medium is a PMA (Phorbol Myristate Acetate)-containing Medium and has been developed for the in vitro cultivation of juvenile melanocytes. Since PMA is a tumor promoting mitogen, it may interfere with some experimental approaches. In these cases, we recommend using the PMA-free Melanocyte Growth Medium M3.While the supplements of the Melanocyte Growth Medium are serum-free, the Basal Medium is manufactured with 0.2% serum.| Media supplement | Final supplement concentration|| Bovine Pituitary Extract | 0.004 ml / ml || Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Insulin (recombinant human) | 5 μg / ml || Hydrocortisone | 0.5 μg / ml || Phorbol Myristate Acetate | 10 ng / ml |
This item is not returnable. View return policy
Catalog No. 50-306-230 Supplier Promocell GmbH Supplier No. C24010
PromoCell Melanocyte Growth Medium is a PMA (Phorbol Myristate Acetate)-containing Medium and has been developed for the in vitro cultivation of juvenile melanocytes. Since PMA is a tumor promoting mitogen, it may interfere with some experimental approaches. In these cases, we recommend using the PMA-free Melanocyte Growth Medium M3.While the supplements of the Melanocyte Growth Medium are serum-free, the Basal Medium is manufactured with 0.2% serum.| Media supplement | Final supplement concentration|| Bovine Pituitary Extract | 0.004 ml / ml || Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Insulin (recombinant human) | 5 μg / ml || Hydrocortisone | 0.5 μg / ml || Phorbol Myristate Acetate | 10 ng / ml |