Learn More
ATCC Melanocyte Growth Kit
Catalog No. 502384882
Encompass
Melanocyte Growth Kit contains components that when added to Dermal Cell Basal Medium (ATCC PCS-200-030) create a complete ATCC Primary Cell Solutions culture environment for melanocytes derived from a normal human epidermis (eg, Primary Epidermal Melanocytes, Normal, Human Neonatal, ATCC PCS-200-012). The complete low serum medium is formulated to support robust melanocyte growth in the absence of cholera toxin and phorbol 12-myristate 13-acetate (PMA).
Catalog No. 50-238-4882 Supplier ATCC Supplier No. PCS200041
Add to Cart
Add to Cart
Melanocyte Growth Kit contains components that when added to Dermal Cell Basal Medium (ATCC PCS-200-030) create a complete ATCC Primary Cell Solutions culture environment for melanocytes derived from a normal human epidermis (eg, Primary Epidermal Melanocytes, Normal, Human Neonatal, ATCC PCS-200-012). The complete low serum medium is formulated to support robust melanocyte growth in the absence of cholera toxin and phorbol 12-myristate 13-acetate (PMA).