Learn More
Medchemexpress LLC HY-P0252 1mg Medchemexpress, α-Melanocyte-Stimulating Hormone (MSH), amide CAS:581-05-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP02521MG
Encompass_Preferred
Medchemexpress, HY-P0252 1mg α-Melanocyte-Stimulating Hormone (MSH), amide CAS:581-05-5 α-Melanocyte-Stimulating Hormone (MSH), amide stimulates melanocortin 1 receptor that results in the activation of adenylyl cyclase . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4080