Learn More
CPC Scientific H-Pro-Leu-Gly-NH2 (trifluoroacetate salt) 50MG
SDP
Supplier:  CPC Scientific MIFI001A
Encompass
This peptide is the C-terminal tripeptide of oxytocin, has been shown to be active in numerous behavioral tests and clinical situations.SEQUENCE: H-Pro-Leu-Gly-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: PLG-NH2MOLECULAR FORMULA: C13H24N4O3MOLECULAR WEIGHT: 284.36STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [2002-44-0]SYNONYMS: melanotropinsRESEARCH AREA: HormonalREFERENCES:S.Bjrkman and H.Sievertsson, Naunyn-Schmiedeberg's Arch. Pharmacol., 298, 79 (1977)T.A.Pugsley and W.Lippmann, Drug Res., 27 II, 2290 (1977)S.Chiu and R.K.Mishra, Eur. J. Pharmacol., 53, 119 (1979)R.Walter et al., Proc. Natl. Acad. Sci. USA, 76, 518 (1979)K.Dolbeare et al., Bioorg. Med. Chem., 11, 4103 (2003)R.S.Khan et al., Int. J. Pept., ID 537639 (2010)SKU(s): MIFI-001A
Catalog No. 50-210-9727