Learn More
Apexbio Technology LLC Melanocyte stimulating hormone release inhibiting factor 2002-44-0 5mg
SDP
Catalog No. 5000096712
Encompass_Preferred
Melanocyte stimulating hormone release inhibiting factor is a small endogenous tripeptide modulator targeting -melanocyte-stimulating hormone ( -MSH) release and dopamine D2/D4 as well as opioid receptors It is designed to inhibit -MSH secretion and modulate dopaminergic and opioid pathways thereby regulating neuropeptide and neurotransmitter signaling Melanocyte stimulating hormone release inhibiting factor exerts its biological activity primarily by allosteric agonism at dopamine D2/D4 receptors and antagonism at opioid receptor sites in addition to inhibiting -MSH release Based on these pharmacological properties melanocyte stimulating hormone release inhibiting factor holds research potential in studies of dopamine signaling opioid pharmacodynamics and neuropeptide regulation
Catalog No. 50-000-96712 Supplier Apexbio Technology LLC Supplier No. A11215
Melanocyte stimulating hormone release inhibiting factor is a small endogenous tripeptide modulator targeting -melanocyte-stimulating hormone ( -MSH) release and dopamine D2/D4 as well as opioid receptors It is designed to inhibit -MSH secretion and modulate dopaminergic and opioid pathways thereby regulating neuropeptide and neurotransmitter signaling Melanocyte stimulating hormone release inhibiting factor exerts its biological activity primarily by allosteric agonism at dopamine D2/D4 receptors and antagonism at opioid receptor sites in addition to inhibiting -MSH release Based on these pharmacological properties melanocyte stimulating hormone release inhibiting factor holds research potential in studies of dopamine signaling opioid pharmacodynamics and neuropeptide regulation