Learn More
Biocare Medical, LLC MELANOMA COCKTAIL 0.5ML
SDP
Catalog No. 5082490
Encompass_Preferred
Melanoma Cocktail is a combination of HMB45 and MART-1, which are coexpressed in the majority of melanomas, as well as solely expressed in certain cases. This cocktail is potentially a sensitive first-order pan melanoma screener.
Catalog No. 50-824-90 Supplier Biocare Medical, LLC Supplier No. CM078B
Add to Cart
Add to Cart
Melanoma Cocktail is a combination of HMB45 and MART-1, which are coexpressed in the majority of melanomas, as well as solely expressed in certain cases. This cocktail is potentially a sensitive first-order pan melanoma screener.