Learn More
Biocare Medical, LLC MELANOMA COCKTAIL 1.0ML
SDP
Supplier:  Biocare Medical, LLC CM078C
Encompass_Preferred
Melanoma Cocktail is a combination of HMB45 and MART-1, which are coexpressed in the majority of melanomas, as well as solely expressed in certain cases. This cocktail is potentially a sensitive first-order pan melanoma screener.
Catalog No. 50-824-91