Learn More
Enzo Life Sciences Melanoma marker (human) monoclonal antibody (HMB45) (Ready-to-Use) (6ml)
SDP
Supplier:  Enzo Life Sciences ENZ30930
Encompass_Preferred
Clone: HMB45. Isotype: IgG1. Immunogen: Human axillary lymph node tissue containing pigmented melanoma metastases. Source: Purified from ascites. Species reactivity: Human. Specificity: Recognizes a human melanoma-specific antigen present in melanocytic cells and junctional nevi. Applications: IHC (PS). Purity: Affinity purified. Formulation: Liquid. Ready-to-use solution in PBS containing 0.1% sodium azide and 1% BSA.
Catalog No. 50-201-2483