Learn More
Medchemexpress LLC HY-N2466 1mg Medchemexpress, Melanotan I CAS:75921-69-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN24661MG
Encompass_Preferred
Medchemexpress, HY-N2466 1mg Melanotan I CAS:75921-69-6 Melanotan I is a synthetic analogue of α-melanocyte stimulating hormone (α-MSH), for gaining a tan. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3929