Learn More
Selleck Chemical LLC Melatonin S1204-50mg
SDP
Supplier:  Selleck Chemical LLC S120450MG
Encompass_Preferred
Melatonin is a MT receptor agonist used as a dietary supplement Melatonin is a selective ATF-6 inhibitor and downregulates COX-2 Melatonin enhances mitophagy and regulates the homeostasis of apoptosis and autophagy
Catalog No. 50-310-8342