Learn More
Apexbio Technology LLC Melatonin(Synonyms: N-acetyl-5-methoxytryptamine, MEL, Circadin, Melovine, Melatol), 10mM (in 1mL DMSO), CAS: 73-31-4.
SDP
Catalog No. 5000060317
Encompass_Preferred
Melatonin (CAS 73-31-4) is a naturally occurring hormone functioning primarily as an agonist at the G protein-coupled melatonin receptors MT and MT Through receptor activation melatonin regulates circadian rhythmicity and contributes significantly to sleep-wake cycle maintenance Additionally melatonin demonstrates immunomodulatory capabilities and possesses robust antioxidant properties in vivo combating oxidative stress Due to these characteristics it is frequently investigated in research related to sleep disorders circadian biology immune function and oxidative stress signaling pathways
Catalog No. 50-000-60317 Supplier Apexbio Technology LLC Supplier No. A28425.1
Melatonin (CAS 73-31-4) is a naturally occurring hormone functioning primarily as an agonist at the G protein-coupled melatonin receptors MT and MT Through receptor activation melatonin regulates circadian rhythmicity and contributes significantly to sleep-wake cycle maintenance Additionally melatonin demonstrates immunomodulatory capabilities and possesses robust antioxidant properties in vivo combating oxidative stress Due to these characteristics it is frequently investigated in research related to sleep disorders circadian biology immune function and oxidative stress signaling pathways