Learn More
Medchemexpress LLC HY-B0075 500mg Medchemexpress, Melatonin CAS:73-31-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0075500MG
Encompass_Preferred
Medchemexpress, HY-B0075 500mg Melatonin CAS:73-31-4 Melatonin is a hormone made by the pineal gland that can activates melatonin receptor. Melatonin plays a role in sleep and possesses important antioxidative and anti-inflammatory properties. Melatonin is a novel selective ATF-6 inhibitor and induces human hepatoma cell apoptosis through COX-2 downregulation[4]. Melatonin attenuates palmitic acid-induced (HY-N0830) mouse granulosa cells apoptosis via endoplasmic reticulum stress[5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2354