Learn More
Medchemexpress LLC Melatonin-d3 (N-Acetyl-5-methoxytryptamine-d3) | 90735-69-6 | 99.1% | C13H13D3N2O2 | 5 MG
SDP
Catalog No. 5000433412
Encompass_Preferred
Melatonin-d3 is the deuterium labeled version of Melatonin. Melatonin is a hormone that activates melatonin receptors, playing a role in sleep and possessing antioxidative and anti-inflammatory properties. It is also identified as a selective ATF-6 inhibitor, inducing apoptosis in human hepatoma cells by downregulating COX-2, and can reduce palmitic acid-induced apoptosis in mouse granulosa cells by alleviating endoplasmic reticulum stress.
  • Used as a tracer
  • Serves as an internal standard for quantitative analysis
  • Stable heavy isotopes for drug development quantitation
  • Deuteration can influence pharmacokinetic and metabolic profiles
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Solvent storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-33412 Supplier Medchemexpress LLC Supplier No. HYB0075S15MG
Melatonin-d3 is the deuterium labeled version of Melatonin. Melatonin is a hormone that activates melatonin receptors, playing a role in sleep and possessing antioxidative and anti-inflammatory properties. It is also identified as a selective ATF-6 inhibitor, inducing apoptosis in human hepatoma cells by downregulating COX-2, and can reduce palmitic acid-induced apoptosis in mouse granulosa cells by alleviating endoplasmic reticulum stress.
  • Used as a tracer
  • Serves as an internal standard for quantitative analysis
  • Stable heavy isotopes for drug development quantitation
  • Deuteration can influence pharmacokinetic and metabolic profiles
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Solvent storage: -80°C for 6 months, -20°C for 1 month