Learn More
Cayman Chemical MelatonIn-d4 500ug
SDP
Supplier:  Cayman Chemical 22291500 UG
Encompass_Preferred
An internal standard for the quantification of melatonin by GC- or LC-MS
Catalog No. 50-298-7791