Learn More
Medchemexpress LLC Melatonin-d4 | 66521-38-8 | 99.0% | C13H12D4N2O2 | 1 MG
SDP
Catalog No. 5000432228
Encompass_Preferred
Melatonin-d4 is deuterium labeled Melatonin, a hormone that activates melatonin receptors. It exhibits antioxidative and anti-inflammatory properties and acts as a selective ATF-6 inhibitor, inducing human hepatoma cell apoptosis through COX-2 downregulation. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated for quantitation during drug development
Catalog No. 50-004-32228 Supplier Medchemexpress LLC Supplier No. HYB0075S1MG
Melatonin-d4 is deuterium labeled Melatonin, a hormone that activates melatonin receptors. It exhibits antioxidative and anti-inflammatory properties and acts as a selective ATF-6 inhibitor, inducing human hepatoma cell apoptosis through COX-2 downregulation. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated for quantitation during drug development