Learn More
Medchemexpress LLC Melatonin-d7 | 615251-68-8 | 98.0% | C13H9D7N2O2 | 1 MG
SDP
Catalog No. 5000432030
Encompass_Preferred
Melatonin-d7 is a deuterium-labeled version of melatonin, a hormone produced by the pineal gland. It activates melatonin receptors and is involved in sleep, exhibiting significant antioxidative and anti-inflammatory properties. Melatonin-d7 also acts as a selective ATF-6 inhibitor, inducing apoptosis in human hepatoma cells by downregulating COX-2, and can reduce palmitic acid-induced apoptosis in mouse granulosa cells through endoplasmic reticulum stress.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-32030 Supplier Medchemexpress LLC Supplier No. HYB0075S21MG
Melatonin-d7 is a deuterium-labeled version of melatonin, a hormone produced by the pineal gland. It activates melatonin receptors and is involved in sleep, exhibiting significant antioxidative and anti-inflammatory properties. Melatonin-d7 also acts as a selective ATF-6 inhibitor, inducing apoptosis in human hepatoma cells by downregulating COX-2, and can reduce palmitic acid-induced apoptosis in mouse granulosa cells through endoplasmic reticulum stress.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs