Learn More
Enzo Life Sciences Melatonin ELISA Kit (1x96wells)
SDP
Catalog No. 502013841
Encompass_Preferred
The Melatonin ELISA kit is a colorimetric competitive immunoassay kit with results in 2 hours. Alternative name: N-acetyl-5-methoxytryptamine. Assay Time: 2 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent .
Catalog No. 50-201-3841 Supplier Enzo Life Sciences Supplier No. ENZKIT1500001
The Melatonin ELISA kit is a colorimetric competitive immunoassay kit with results in 2 hours. Alternative name: N-acetyl-5-methoxytryptamine. Assay Time: 2 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent .