Learn More
Abcam Melatonin ELISA Kit, 96TEST
Catalog No. 502654931
Encompass_Preferred
Melatonin ELISA Kit is a Competitive ELISA kit for the measurement of Melatonin in Plasma, Tissue Homogenate, Serum, Other biological fluids samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-4931 Supplier Abcam Supplier No. AB28525196TEST
Melatonin ELISA Kit is a Competitive ELISA kit for the measurement of Melatonin in Plasma, Tissue Homogenate, Serum, Other biological fluids samples.
The product is subject to the following: Abcam Restricted Use Statement