Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedical™ Melatonin ELISA Kit - Serum

Catalog No. mp07p534a Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
96 Wells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP07P534A 96 Wells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP07P534A Supplier MP Biomedical™ Supplier No. 07P534A
Only null left
Add to Cart
Add to Cart

Quantitative Enzyme-linked Immunosorbent Assay

The Melatonin ELISA kit (serum) is intended for the measurement of melatonin in human serum.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Kit and Assay
Sample Type Serum
Target Species Human
Quantity 96 Wells
Regulatory Status For research use only (RUO)
Sample Volume 25 μL
Storage Requirements 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.