Learn More
Alpco Ltd Melatonin Extraction Columns
SDP
Catalog No. NC2284623
Encompass
For use with 12-RK-MEL2-U. C18 solid phase extraction columns available in boxes of 10¿0 units for extraction from biological samples. Each extraction column can be used up to five times according to the extraction procedures described in the Melatonin RIA Kit (12-RK-MEL2-U) instructions for use.
This item is not returnable. View return policy
Catalog No. NC2284623 Supplier Alpco Ltd Supplier No. 12BMECX
For use with 12-RK-MEL2-U. C18 solid phase extraction columns available in boxes of 10¿0 units for extraction from biological samples. Each extraction column can be used up to five times according to the extraction procedures described in the Melatonin RIA Kit (12-RK-MEL2-U) instructions for use.