Learn More
TRIBIOSCIENCE INC Melatonin Monoclonal Antibody, 1MG
SDP
Catalog No. NC3496709
Encompass
This monoclonal antibody is highly specific to melatonin and is generated through the hybridization of P3x63-Ag 9.653 myeloma cells with spleen cells from a Balb/c mouse. Classified as an IgG1 isotype with a kappa light chain, it is sourced from mouse ascites and purified using DEAE chromatography to achieve a purity greater than 95%, as confirmed by HPLC and SDS-PAGE. The antibody is provided in a purified liquid format at a concentration of 5.35 mg/ml (OD280 nm, E0.1% = 1.40) and is unconjugated. It is designed for use in melatonin quantitative assays via EIA.
Catalog No. NC3496709 Supplier TRIBIOSCIENCE INC Supplier No. TBS101571MG
This monoclonal antibody is highly specific to melatonin and is generated through the hybridization of P3x63-Ag 9.653 myeloma cells with spleen cells from a Balb/c mouse. Classified as an IgG1 isotype with a kappa light chain, it is sourced from mouse ascites and purified using DEAE chromatography to achieve a purity greater than 95%, as confirmed by HPLC and SDS-PAGE. The antibody is provided in a purified liquid format at a concentration of 5.35 mg/ml (OD280 nm, E0.1% = 1.40) and is unconjugated. It is designed for use in melatonin quantitative assays via EIA.