Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melatonin, MP Biomedicals

Catalog No. MP021022541 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021022541 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021022541 Supplier MP Biomedicals, Inc Supplier No. 02102254.1
Add to Cart
Add to Cart

  • Melatonin is a hormone of the pineal gland

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-31-4
Molecular Formula C13H16N2O2
Molecular Weight (g/mol) 232.283
MDL Number MFCD00005655
InChI Key DRLFMBDRBRZALE-UHFFFAOYSA-N
Synonym melatonin, melatonine, n-acetyl-5-methoxytryptamine, circadin, 5-methoxy-n-acetyltryptamine, n-2-5-methoxy-1h-indol-3-yl ethyl acetamide, melatol, melatonex, melovine
PubChem CID 896
ChEBI CHEBI:16796
IUPAC Name N-[2-(5-methoxy-1H-indol-3-yl)ethyl]acetamide
SMILES CC(=O)NCCC1=CNC2=C1C=C(C=C2)OC

Specifications

Melting Point 114°C to 124°C
Color White
Linear Formula C13H16N2O2
Quantity 100 mg
Solubility Information Soluble in methanol, ethanol (50mg/mL - clear to slightly hazy, colorless to faint yellow solution); Very slightly soluble in 0.1 N HCl (0.8mg/mL), 0.1 N NaOH (0.2mg/mL) or water (0.1mg/mL).
Formula Weight 232.28
Physical Form Powder
Chemical Name or Material MELATONIN
Recommended Storage Store at -0°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.