Learn More
Biomatik Corporation Melatonin (MT) CLIA Kit
Supplier:  Biomatik Corporation EKU12346
Encompass_Preferred
Melatonin (MT) CLIA Kit for detecting MT in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 3.9-1,000pg/ml. Sensitivity: 1.7pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Target Synonyms: N-Acetyl-5-Methoxytryptamine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6952