Learn More
Biomatik Corporation Melatonin (MT) Instant ELISA Kit
Supplier:  Biomatik Corporation EKU10367
Encompass_Preferred
Melatonin (MT) Instant ELISA Kit for detecting MT in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 6.17-500pg/ml. Sensitivity: 2.51pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 1h, 10min. Target Synonyms: N-Acetyl-5-Methoxytryptamine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5433