Learn More
Cayman Chemical MelatonIn Receptr TYP 1Ah 10ug
SDP
Catalog No. 502932999
Encompass_Preferred
Source: Human MT1 purified from a baculovirus overexpression system in Sf9 cells • MW = 44.8 kDa
Catalog No. 50-293-2999 Supplier Cayman Chemical Supplier No. 2473110 UG
Source: Human MT1 purified from a baculovirus overexpression system in Sf9 cells • MW = 44.8 kDa