Learn More
Alpco Ltd Melatonin RIA
SDP
Supplier:  Alpco Ltd 12RKMEL2U
Encompass
The Melatonin RIA kit is intended for the quantitative determination of melatonin (1-3) in serum, plasma, urine and other biological specimen upon C18 solid phase extraction. Research Use Only. Not for Use in Diagnostic Procedures. Recommend to use 12-B-MECX Melatonin Extraction Columns.
Catalog No. NC2284544