Learn More
Eagle Biosciences Inc Melatonin Saliva ELISA
SDP
Supplier:  Eagle Biosciences Inc MEL32K01
Encompass
The Eagle Biosciences Melatonin Saliva ELISA Assay Kit is a rapid, sensitive and specific EIA designed and validated for the Direct quantitative measurement of Melatonin in human saliva.
Catalog No. 50-187-8679