Learn More
Medchemexpress LLC Melatonin (standard) | 73-31-4 | 99.27% | C13H16N2O2 | 250 MG
SDP
Catalog No. 5000358506
Encompass_Preferred
Melatonin (Standard) is an analytical standard of Melatonin, primarily intended for research and analytical applications. This hormone, produced by the pineal gland, activates melatonin receptors and plays a crucial role in sleep. It also exhibits important antioxidative and anti-inflammatory properties, making it valuable for various scientific studies. The compound has a molecular weight of 232.28 and appears as an off-white to light yellow solid.
  • Functions as a novel selective ATF-6 inhibitor
  • Induces human hepatoma cell apoptosis through COX-2 downregulation
  • Attenuates palmitic acid-induced mouse granulosa cell apoptosis via endoplasmic reticulum stress
  • Suitable for qualitative, quantitative, and methodological research experiments
  • Used in applications such as HPLC, GC, and MS
Catalog No. 50-003-58506 Supplier Medchemexpress LLC Supplier No. 250MGHYB0075R
Melatonin (Standard) is an analytical standard of Melatonin, primarily intended for research and analytical applications. This hormone, produced by the pineal gland, activates melatonin receptors and plays a crucial role in sleep. It also exhibits important antioxidative and anti-inflammatory properties, making it valuable for various scientific studies. The compound has a molecular weight of 232.28 and appears as an off-white to light yellow solid.
  • Functions as a novel selective ATF-6 inhibitor
  • Induces human hepatoma cell apoptosis through COX-2 downregulation
  • Attenuates palmitic acid-induced mouse granulosa cell apoptosis via endoplasmic reticulum stress
  • Suitable for qualitative, quantitative, and methodological research experiments
  • Used in applications such as HPLC, GC, and MS