Learn More
AdipoGen Meleagrin (1 mg)
SDP
Supplier:  AdipoGen AGCN20451M001
Encompass_Preferred
Chemical. CAS: 71751-77-4. Formula: C23H23N5O4. MW: 433.5. Isolated from Penicillium sp. Mycotoxin. Alkaloid antibiotic. Bacterial enoyl-acyl carrier protein reductase (FabI) inhibitor. Antineoplastic activity. Moderate cytotoxicity against A-549 and HL-60 cell lines. Induces cell cycle arrest through G2/M phase, presumably by inhibiting tubulin polymerization. Antifouling agent against the barnacle Balanus amphitrite.
Catalog No. 50-205-3619