Learn More
Medchemexpress LLC Melevodopa hydrochloride | 1421-65-4 | 99.8% | 247.68 | 5 G
SDP
Catalog No. 5000453959
Encompass_Preferred
Melevodopa hydrochloride is an effervescent Levodopa prodrug. Levodopa is an orally active metabolic precursor of the neurotransmitter dopamine and has anti-allodynic effects and potential for treating Parkinson's disease.
  • Effervescent prodrug of Levodopa
  • Metabolic precursor of dopamine
  • Anti-allodynic effects
  • Potential for Parkinson's disease treatment
Catalog No. 50-004-53959 Supplier Medchemexpress LLC Supplier No. HYW0119635G
Melevodopa hydrochloride is an effervescent Levodopa prodrug. Levodopa is an orally active metabolic precursor of the neurotransmitter dopamine and has anti-allodynic effects and potential for treating Parkinson's disease.
  • Effervescent prodrug of Levodopa
  • Metabolic precursor of dopamine
  • Anti-allodynic effects
  • Potential for Parkinson's disease treatment