Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Melflufen (hydrochloride) | 380449-54-7 | 98.1% | 534.88 | C24H31Cl3FN3O3 | 25 MG
SDP

Catalog No. 5000244209
Encompass_Preferred

Melflufen hydrochloride is a peptide-conjugated alkylating prodrug of melphalan supplied as a hydrochloride salt for research use. It demonstrates antitumor activity in preclinical studies, is provided as a high-purity solid for in vitro and biochemical oncology research, and should be handled according to the manufacturer safety data sheet.

  • Peptide-conjugated alkylating prodrug.
  • Demonstrates antitumor activity against multiple myeloma cells.
  • Off-white to light yellow solid appearance.
  • Purity approximately 98.06%.
  • Molecular weight 534.88 g/mol.
  • Available in small milligram pack sizes for research use.
  • Store sealed at 4°C and protect from moisture; follow SDS for details.

Catalog No. 50-002-44209 Supplier Medchemexpress LLC Supplier No. HY105019A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Melflufen hydrochloride is a peptide-conjugated alkylating prodrug of melphalan supplied as a hydrochloride salt for research use. It demonstrates antitumor activity in preclinical studies, is provided as a high-purity solid for in vitro and biochemical oncology research, and should be handled according to the manufacturer safety data sheet.

  • Peptide-conjugated alkylating prodrug.
  • Demonstrates antitumor activity against multiple myeloma cells.
  • Off-white to light yellow solid appearance.
  • Purity approximately 98.06%.
  • Molecular weight 534.88 g/mol.
  • Available in small milligram pack sizes for research use.
  • Store sealed at 4°C and protect from moisture; follow SDS for details.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.