Learn More
Apexbio Technology LLC Melibiose 585-99-9 500mg
SDP
Catalog No. 5000091990
Encompass_Preferred
Melibiose (CAS 585-99-9) is a disaccharide composed of galactose and glucose linked via an alpha(1-6) glycosidic bond Studies have demonstrated that melibiose can modulate specific signaling pathways and enzymatic activities in vitro indicating its capacity to influence various biological processes Owing to these properties melibiose is utilized as a tool compound in research settings for investigating disease models and elucidating biological functions relevant to its pathway modulation
Catalog No. 50-000-91990 Supplier Apexbio Technology LLC Supplier No. B7901500
Melibiose (CAS 585-99-9) is a disaccharide composed of galactose and glucose linked via an alpha(1-6) glycosidic bond Studies have demonstrated that melibiose can modulate specific signaling pathways and enzymatic activities in vitro indicating its capacity to influence various biological processes Owing to these properties melibiose is utilized as a tool compound in research settings for investigating disease models and elucidating biological functions relevant to its pathway modulation